Search
Search
| Gene Symbol: Snupn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004270.1 | NP_001004270.1 | 4 - 5 | 532 | 76 | |
| XM_006243124.1 | XP_006243186.1 | 5 - 6 | 1158 | 76 | ||
| XM_006243125.1 | XP_006243187.1 | 5 - 6 | 819 | 76 | ||
| XM_006243126.1 | XP_006243188.1 | 5 - 6 | 838 | 76 | ||
| XM_006243127.2 | XP_006243189.1 | 5 - 6 | 782 | 76 | ||
| XM_006243128.2 | XP_006243190.1 | 5 - 6 | 1158 | 76 | ||
| GenBank mRNA | BC079451.1 | - | 4 - 5 | 532 | 7121061 | 76 |
Set Membership: |
|