Search
Search
| Gene Symbol: Uaca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195564.1 | NP_001182493.1 | 4 - 5 | 428 | 66 | |
| XM_006243205.2 | XP_006243267.1 | 4 - 5 | 484 | 66 | ||
| XM_006243206.1 | XP_006243268.1 | 4 - 5 | 628 | 66 | ||
| XM_008766274.1 | XP_008764496.1 | 4 - 5 | 484 | 66 | ||
| GenBank mRNA | FQ216984.1 | - | 4 - 5 | 507 | 66 | |
Set Membership: |
|