Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01418038_m1
          See other LOXL1 GE Assays ›
          Gene Symbol:
          Loxl1
          Gene Name:
          lysyl oxidase-like 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 63067757 - 63092124 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01418038_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Loxl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012125.1 NP_001012125.1 1 - 2 1516 82
          GenBank mRNA BC089224.1 - 1 - 2 1516 7309311 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002650.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Loxl1

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to lipopolysaccharide
          aorta development
          oxidation-reduction process
          molecular_function
          copper ion binding
          oxidoreductase activity, acting on the CH-NH2 group of donors, oxygen as acceptor

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P19907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609151 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76639 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59725 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19907 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19907 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P19907 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19907 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P546338 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10997 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23182 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10917890 - -
          Affymetrix Rat Gene 1.0 ST Array 10917886 - -
          Affymetrix Rat Gene 1.0 ST Array 10917887 - -
          Affymetrix Rat Gene 1.0 ST Array 10917888 - -
          Affymetrix Rat Gene 1.0 ST Array 10917885 - -
          Affymetrix Rat Gene 1.0 ST Array 10917891 - -
          Affymetrix Rat Gene 1.0 ST Array 10917889 - -
          Affymetrix Rat Gene 1.0 ST Array 10917884 - -
          Affymetrix Rat Gene 1.0 ST Array 10917892 - -
          Affymetrix Rat Gene 1.1 ST Array 10917883 - -
          Rat Genome 230 2.0 Array 1388902_at - -
          Rat Genome 230 2.0 Array 1388902_PM_at - -
          Affymetrix HT Rat Focus Array 1388902_at - -
          Affymetrix HT Rat Focus Array 1388902_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800021549.rn.1 TC0800002650.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021547.rn.1 TC0800002650.rn.1 RTA TC
          Rat Expression Set 230 1388902_at - -
          Rat Expression Set 230 1388902_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10917883 - -
          Rat Genome U34 Set rc_AA859805_at - -
          Rat Genome U34 Set rc_AA800844_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530405 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376846 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline