Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01418372_m1
          See other WDR61 GE Assays ›
          Gene Symbol:
          Wdr61
          Gene Name:
          WD repeat domain 61
          Gene Aliases:
          RGD1308228
          Chromosome Location:
          Chr.8: 59296339 - 59313626 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01418372_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wdr61
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025743.1 NP_001020914.1 9 - 10 850 74
          XM_006243085.2 XP_006243147.1 9 - 10 851 74
          XM_006243086.2 XP_006243148.1 9 - 10 904 74
          XM_006243087.2 XP_006243149.1 9 - 10 877 74
          GenBank mRNA BC098059.1 - 9 - 10 850 7458076 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wdr61

          Gene Ontology Categories:

          Function(s)

          transcription, DNA-templated
          Wnt signaling pathway
          positive regulation of transcription elongation from RNA polymerase II promoter
          negative regulation of myeloid cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of histone H3-K4 methylation
          histone H3-K4 trimethylation
          positive regulation of histone H3-K79 methylation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P537715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37026 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40681 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P537715 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P537715 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P537715 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P537715 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34305 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44170 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10917586 - -
          Affymetrix Rat Gene 1.0 ST Array 10917591 - -
          Affymetrix Rat Gene 1.0 ST Array 10917589 - -
          Affymetrix Rat Gene 1.0 ST Array 10917584 - -
          Affymetrix Rat Gene 1.0 ST Array 10917587 - -
          Affymetrix Rat Gene 1.0 ST Array 10917588 - -
          Affymetrix Rat Gene 1.0 ST Array 10917590 - -
          Affymetrix Rat Gene 1.0 ST Array 10917585 - -
          Affymetrix Rat Gene 1.1 ST Array 10917583 - -
          Rat Genome 230 2.0 Array 1389323_PM_at - -
          Rat Genome 230 2.0 Array 1389323_at - -
          Affymetrix HT Rat Focus Array 1389323_PM_at - -
          Affymetrix HT Rat Focus Array 1389323_at - -
          Affymetrix Rat Clariom S Assay PSR0800020882.rn.1 TC0800002584.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020883.rn.1 TC0800002584.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020884.rn.1 TC0800002584.rn.1 RTA TC
          Rat Expression Set 230 1389323_PM_at - -
          Rat Expression Set 230 1389323_at - -
          Affymetrix Rat Gene 1.0 ST Array 10917583 - -
          Rat Genome U34 Set rc_AA893626_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21095237 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364741 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline