Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01418496_m1
          See other DDX6 GE Assays ›
          Gene Symbol:
          Ddx6
          Gene Name:
          DEAD (Asp-Glu-Ala-Asp) box helicase 6
          Gene Aliases:
          RGD1564560
          Chromosome Location:
          Chr.8: 48924768 - 48961394 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01418496_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddx6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109292.1 NP_001102762.1 7 - 8 1088 69
          XM_006242950.2 XP_006243012.1 7 - 8 1088 69
          XM_006242951.2 XP_006243013.1 7 - 8 1102 69
          XM_006242952.2 XP_006243014.1 7 - 8 1097 69
          XM_006242954.2 XP_006243016.1 7 - 8 1371 69

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          cytoplasmic mRNA processing body assembly
          helicase activity
          ATP binding
          protein domain specific binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P649177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475780 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136015 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38956 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P649177 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P649177 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P649177 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P335165 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P793224 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P649177 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32134 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10909545 - -
          Affymetrix Rat Gene 1.0 ST Array 10915784 - -
          Affymetrix Rat Gene 1.0 ST Array 10763429 - -
          Affymetrix Rat Gene 1.0 ST Array 10909542 - -
          Affymetrix Rat Gene 1.0 ST Array 10909548 - -
          Affymetrix Rat Gene 1.0 ST Array 10909537 - -
          Affymetrix Rat Gene 1.0 ST Array 10909546 - -
          Affymetrix Rat Gene 1.0 ST Array 10909547 - -
          Affymetrix Rat Gene 1.0 ST Array 10909540 - -
          Affymetrix Rat Gene 1.0 ST Array 10909538 - -
          Affymetrix Rat Gene 1.0 ST Array 10909544 - -
          Affymetrix Rat Gene 1.0 ST Array 10909539 - -
          Affymetrix Rat Gene 1.0 ST Array 10909543 - -
          Affymetrix Rat Gene 1.0 ST Array 10909541 - -
          Affymetrix Rat Gene 1.1 ST Array 10763428 - -
          Affymetrix Rat Gene 1.1 ST Array 10909536 - -
          Affymetrix Rat Gene 1.1 ST Array 10915783 - -
          Rat Genome 230 2.0 Array 1389868_PM_at - -
          Rat Genome 230 2.0 Array 1389868_at - -
          Rat Genome 230 2.0 Array 1392854_at - -
          Rat Genome 230 2.0 Array 1392854_PM_at - -
          Affymetrix HT Rat Focus Array 1389868_at - -
          Affymetrix HT Rat Focus Array 1389868_PM_at - -
          Affymetrix HT Rat Focus Array 1392854_at - -
          Affymetrix HT Rat Focus Array 1392854_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800003668.rn.1 TC0800000573.rn.1 RTA TC
          Rat Expression Set 230 1389868_PM_at - -
          Rat Expression Set 230 1389868_at - -
          Rat Expression Set 230 1392854_PM_at - -
          Rat Expression Set 230 1392854_at - -
          Affymetrix Rat Gene 1.0 ST Array 10915783 - -
          Affymetrix Rat Gene 1.0 ST Array 10909536 - -
          Affymetrix Rat Gene 1.0 ST Array 10763428 - -
          Rat Genome U34 Set rc_AA850780_at - -
          Rat Genome U34 Set rc_AA925853_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21989916 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351983 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline