Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01418560_g1
          See other SCN2B GE Assays ›
          Gene Symbol:
          Scn2b
          Gene Name:
          sodium channel, voltage-gated, type II, beta subunit
          Gene Aliases:
          SCNB2
          Chromosome Location:
          Chr.8: 49419003 - 49431110 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01418560_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scn2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012877.1 NP_037009.1 2 - 3 404 70
          XM_006242895.2 XP_006242957.1 2 - 3 356 70
          GenBank mRNA U37026.1 - 2 - 3 404 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000592.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scn2b

          Panther Classification:

          Molecular Function -

          myelin protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          sodium ion transmembrane transport
          response to pyrethroid
          cardiac muscle contraction
          regulation of atrial cardiac muscle cell membrane depolarization
          cardiac muscle cell action potential involved in contraction
          membrane depolarization during cardiac muscle cell action potential
          regulation of heart rate by cardiac conduction
          regulation of sodium ion transmembrane transporter activity
          voltage-gated sodium channel activity
          protein binding
          sodium channel regulator activity
          voltage-gated sodium channel activity involved in cardiac muscle cell action potential

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P121991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693219 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142952 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P121991 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P121991 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P121991 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P597619 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21579 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10909620 - -
          Affymetrix Rat Gene 1.0 ST Array 10909619 - -
          Affymetrix Rat Gene 1.0 ST Array 10909618 - -
          Affymetrix Rat Gene 1.0 ST Array 10909617 - -
          Affymetrix Rat Gene 1.1 ST Array 10909616 - -
          Rat Genome 230 2.0 Array 1369311_PM_at - -
          Rat Genome 230 2.0 Array 1369311_at - -
          Affymetrix HT Rat Focus Array 1369311_at - -
          Affymetrix HT Rat Focus Array 1369311_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800003816.rn.1 TC0800000592.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800003818.rn.1 TC0800000592.rn.1 RTA TC
          Rat Expression Set 230 1369311_at - -
          Rat Expression Set 230 1369311_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10909616 - -
          Rat Genome U34 Set U37147_at - -
          Rat Genome U34 Set U37147_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21967707 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367526 - -
          RatRef-12 Expression BeadChip ILMN_50963 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline