Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01418692_m1
          See other LPIN2 GE Assays ›
          Gene Symbol:
          Lpin2
          Gene Name:
          lipin 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 119517101 - 119591533 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01418692_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lpin2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108236.1 NP_001101706.1 12 - 13 1801 71
          XM_006245662.2 XP_006245724.1 12 - 13 4105 71
          XM_006245663.2 XP_006245725.1 12 - 13 5122 71
          XM_006245664.2 XP_006245726.1 12 - 13 2350 71
          XM_006245665.2 XP_006245727.1 12 - 13 4107 71
          XM_006245666.2 XP_006245728.1 12 - 13 1839 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001343.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          fatty acid catabolic process
          dephosphorylation
          triglyceride biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          transcription coactivator activity
          phosphatidate phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1045748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271754 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31688 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169236 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135632 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27648 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150320 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30604 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20852 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57747 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49303 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60225 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62231 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1045748 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1045748 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1045748 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1045748 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45595 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64423 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56784 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10926042 - -
          Affymetrix Rat Gene 1.0 ST Array 10926037 - -
          Affymetrix Rat Gene 1.0 ST Array 10926046 - -
          Affymetrix Rat Gene 1.0 ST Array 10926038 - -
          Affymetrix Rat Gene 1.0 ST Array 10926049 - -
          Affymetrix Rat Gene 1.0 ST Array 10926040 - -
          Affymetrix Rat Gene 1.0 ST Array 10926047 - -
          Affymetrix Rat Gene 1.0 ST Array 10926036 - -
          Affymetrix Rat Gene 1.0 ST Array 10926053 - -
          Affymetrix Rat Gene 1.0 ST Array 10926051 - -
          Affymetrix Rat Gene 1.0 ST Array 10926052 - -
          Affymetrix Rat Gene 1.0 ST Array 10926044 - -
          Affymetrix Rat Gene 1.0 ST Array 10926045 - -
          Affymetrix Rat Gene 1.0 ST Array 10926041 - -
          Affymetrix Rat Gene 1.0 ST Array 10926039 - -
          Affymetrix Rat Gene 1.0 ST Array 10926050 - -
          Affymetrix Rat Gene 1.0 ST Array 10926043 - -
          Affymetrix Rat Gene 1.0 ST Array 10926048 - -
          Affymetrix Rat Gene 1.0 ST Array 10926035 - -
          Affymetrix Rat Gene 1.1 ST Array 10926029 - -
          Rat Genome 230 2.0 Array 1396398_PM_at - -
          Rat Genome 230 2.0 Array 1383665_PM_at - -
          Rat Genome 230 2.0 Array 1383665_at - -
          Rat Genome 230 2.0 Array 1396398_at - -
          Affymetrix HT Rat Focus Array 1396398_at - -
          Affymetrix HT Rat Focus Array 1396398_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900011367.rn.1 TC0900001343.rn.1 RTA TC
          Rat Expression Set 230 1383665_at - -
          Rat Expression Set 230 1383665_PM_at - -
          Rat Expression Set 230 1396398_PM_at - -
          Rat Expression Set 230 1396398_at - -
          Affymetrix Rat Gene 1.0 ST Array 10926029 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21887984 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356179 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline