Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01418715_m1
          See other TGIF1 GE Assays ›
          Gene Symbol:
          Tgif1
          Gene Name:
          TGFB-induced factor homeobox 1
          Gene Aliases:
          Tfig, Tgif
          Chromosome Location:
          Chr.9: 119181079 - 119190698 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01418715_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgif1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001015020.1 NP_001015020.1 2 - 3 328 84
          GenBank mRNA BC091264.1 - 2 - 3 328 7379061 84
          BC127461.1 - 2 - 3 416 7389058 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002675.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tgif1

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          neural tube closure
          determination of left/right symmetry
          negative regulation of cell proliferation
          dorsal/ventral pattern formation
          regulation of gastrulation
          negative regulation of gene expression
          nodal signaling pathway
          response to drug
          positive regulation of neuron differentiation
          positive regulation of fibroblast proliferation
          negative regulation of retinoic acid receptor signaling pathway
          retina development in camera-type eye
          cellular response to growth factor stimulus
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          chromatin binding
          co-SMAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P044435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153925 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75682 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044435 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044435 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P044435 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P157222 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29613 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39809 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10930412 - -
          Affymetrix Rat Gene 1.0 ST Array 10930413 - -
          Affymetrix Rat Gene 1.0 ST Array 10930414 - -
          Affymetrix Rat Gene 1.0 ST Array 10930415 - -
          Affymetrix Rat Gene 1.1 ST Array 10930411 - -
          Rat Genome 230 2.0 Array 1373421_PM_at - -
          Rat Genome 230 2.0 Array 1373421_at - -
          Affymetrix HT Rat Focus Array 1373421_PM_at - -
          Affymetrix HT Rat Focus Array 1373421_at - -
          Affymetrix Rat Clariom S Assay PSR0900021642.rn.1 TC0900002675.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900021643.rn.1 TC0900002675.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900021648.rn.1 TC0900002675.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900021649.rn.1 TC0900002675.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900021640.rn.1 TC0900002675.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900021638.rn.1 TC0900002675.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900021639.rn.1 TC0900002675.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900021641.rn.1 TC0900002675.rn.1 RTA TC
          Rat Expression Set 230 1373421_PM_at - -
          Rat Expression Set 230 1373421_at - -
          Affymetrix Rat Gene 1.0 ST Array 10930411 - -
          Rat Genome U34 Set rc_AI178025_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22351718 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651152 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline