Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01418781_g1
          See other MYL12A GE Assays ›
          Gene Symbol:
          Myl12a
          Gene Name:
          myosin, light chain 12A, regulatory, non-sarcomeric
          Gene Aliases:
          RGD1309537, Rlc-a
          Chromosome Location:
          Chr.9: 119325282 - 119332967 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01418781_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myl12a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135017.1 NP_001128489.1 2 - 3 316 65
          XM_006245667.2 XP_006245729.1 2 - 3 470 65
          XM_006245668.2 XP_006245730.1 2 - 3 411 65
          GenBank mRNA EV767772.1 - 346 65
          FQ215121.1 - 2 - 3 271 65
          FQ215776.1 - 460 65
          FQ217399.1 - 2 - 3 362 65
          FQ217558.1 - 2 - 3 246 65
          FQ220614.1 - 302 65
          FQ223132.1 - 2 - 3 280 65
          FQ228530.1 - 2 - 3 273 65
          FQ229140.1 - 2 - 3 525 65
          FQ232936.1 - 2 - 3 252 65
          X05566.1 - 2 - 3 258 65
          Z32519.1 - 2 - 3 258 65

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell shape
          platelet aggregation
          protein targeting to plasma membrane
          calcium ion binding
          glutamate receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P105231 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P105228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617826 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176221 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105228 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105231 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105231 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105228 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P105228 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P105231 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P226478 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P160321 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26911 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10930423 - -
          Affymetrix Rat Gene 1.0 ST Array 10930424 - -
          Affymetrix Rat Gene 1.0 ST Array 10930422 - -
          Affymetrix Rat Gene 1.1 ST Array 10930421 - -
          Rat Genome 230 2.0 Array 1388114_at - -
          Rat Genome 230 2.0 Array 1388114_PM_at - -
          Affymetrix HT Rat Focus Array 1388114_PM_at - -
          Affymetrix HT Rat Focus Array 1388114_at - -
          Affymetrix Rat Clariom S Assay PSR0900022602.rn.1 TC0900002726.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900022599.rn.1 TC0900002726.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900022604.rn.1 TC0900002726.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900022605.rn.1 TC0900002726.rn.1 RTA TC
          Rat Expression Set 230 1388114_PM_at - -
          Rat Expression Set 230 1388114_at - -
          Affymetrix Rat Gene 1.0 ST Array 10930421 - -
          Rat Genome U34 Set X05566_i_at - -
          Rat Genome U34 Set X54617mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353763 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline