Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01419027_m1
          See other POLA1 GE Assays ›
          Gene Symbol:
          Pola1
          Gene Name:
          polymerase (DNA directed), alpha 1, catalytic subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 62382604 - 62698830 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01419027_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pola1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053479.1 NP_445931.1 23 - 24 2697 68
          GenBank mRNA AJ011605.1 - 23 - 24 2697 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001857.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA synthesis involved in DNA repair
          DNA replication
          DNA replication, synthesis of RNA primer
          DNA replication initiation
          DNA strand elongation involved in DNA replication
          leading strand elongation
          lagging strand elongation
          DNA repair
          double-strand break repair via nonhomologous end joining
          cell proliferation
          DNA biosynthetic process
          nucleotide binding
          nucleoside binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          DNA-directed DNA polymerase activity
          DNA primase activity
          purine nucleotide binding
          pyrimidine nucleotide binding
          protein kinase binding
          metal ion binding
          protein heterodimerization activity
          4 iron, 4 sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15924 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15924 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P231678 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P231678 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15924 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15924 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P231678 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15924 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P368241 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27831 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55754 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75477 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75963 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27479 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46090 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68381 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42106 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38533 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34490 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79148 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91258 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10327 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24681 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10938367 - -
          Affymetrix Rat Gene 1.0 ST Array 10938350 - -
          Affymetrix Rat Gene 1.0 ST Array 10938382 - -
          Affymetrix Rat Gene 1.0 ST Array 10938348 - -
          Affymetrix Rat Gene 1.0 ST Array 10938361 - -
          Affymetrix Rat Gene 1.0 ST Array 10938365 - -
          Affymetrix Rat Gene 1.0 ST Array 10938356 - -
          Affymetrix Rat Gene 1.0 ST Array 10938381 - -
          Affymetrix Rat Gene 1.0 ST Array 10938352 - -
          Affymetrix Rat Gene 1.0 ST Array 10938357 - -
          Affymetrix Rat Gene 1.0 ST Array 10938363 - -
          Affymetrix Rat Gene 1.0 ST Array 10938360 - -
          Affymetrix Rat Gene 1.0 ST Array 10938347 - -
          Affymetrix Rat Gene 1.0 ST Array 10938371 - -
          Affymetrix Rat Gene 1.0 ST Array 10938377 - -
          Affymetrix Rat Gene 1.0 ST Array 10938370 - -
          Affymetrix Rat Gene 1.0 ST Array 10938375 - -
          Affymetrix Rat Gene 1.0 ST Array 10938353 - -
          Affymetrix Rat Gene 1.0 ST Array 10938379 - -
          Affymetrix Rat Gene 1.0 ST Array 10938383 - -
          Affymetrix Rat Gene 1.0 ST Array 10938372 - -
          Affymetrix Rat Gene 1.0 ST Array 10938368 - -
          Affymetrix Rat Gene 1.0 ST Array 10938349 - -
          Affymetrix Rat Gene 1.0 ST Array 10938359 - -
          Affymetrix Rat Gene 1.0 ST Array 10938374 - -
          Affymetrix Rat Gene 1.0 ST Array 10938351 - -
          Affymetrix Rat Gene 1.0 ST Array 10938355 - -
          Affymetrix Rat Gene 1.0 ST Array 10938380 - -
          Affymetrix Rat Gene 1.0 ST Array 10938373 - -
          Affymetrix Rat Gene 1.0 ST Array 10938354 - -
          Affymetrix Rat Gene 1.0 ST Array 10938358 - -
          Affymetrix Rat Gene 1.0 ST Array 10938366 - -
          Affymetrix Rat Gene 1.0 ST Array 10938362 - -
          Affymetrix Rat Gene 1.0 ST Array 10938364 - -
          Affymetrix Rat Gene 1.0 ST Array 10938378 - -
          Affymetrix Rat Gene 1.1 ST Array 10938346 - -
          Rat Genome 230 2.0 Array 1376611_PM_at - -
          Rat Genome 230 2.0 Array 1376611_at - -
          Affymetrix HT Rat Focus Array 1376611_PM_at - -
          Affymetrix HT Rat Focus Array 1376611_at - -
          Affymetrix Rat Clariom S Assay PSR0X00011692.rn.1 TC0X00001857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00011694.rn.1 TC0X00001857.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00011693.rn.1 TC0X00001857.rn.1 RTA TC
          Rat Expression Set 230 1376611_PM_at - -
          Rat Expression Set 230 1376611_at - -
          Affymetrix Rat Gene 1.0 ST Array 10938346 - -
          Rat Genome U34 Set AJ011605_at - -
          Rat Genome U34 Set rc_AI103682_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353900 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362800 - -
          RatRef-12 Expression BeadChip ILMN_55699 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0