Search
Search

Itgb1bp2| Gene Symbol: Itgb1bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108245.2 | NP_001101715.1 | 4 - 5 | 371 | 129 | |
| GenBank mRNA | FM116382.1 | - | 4 - 5 | 330 | 129 | |
| FM118664.1 | - | 4 - 5 | 355 | 129 | ||
| FQ215476.1 | - | 4 - 5 | 366 | 129 | ||
| FQ216044.1 | - | 4 - 5 | 370 | 129 | ||
| FQ216100.1 | - | 4 - 5 | 347 | 129 | ||
| FQ216185.1 | - | 4 - 5 | 367 | 129 | ||
| FQ224925.1 | - | 4 - 5 | 368 | 129 | ||
Set Membership: |
|