Search
Search

Magebl1| Gene Symbol: Magebl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128493.1 | NP_001121965.1 | 3 | 784 | 152 | |
| XM_006257011.2 | XP_006257073.1 | 3 | 780 | 152 | ||
| GenBank mRNA | BC158628.1 | - | 3 | 784 | 7455432 | 152 |
Set Membership: |
|