Search
Search

RGD1562161| Gene Symbol: RGD1562161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127568.1 | NP_001121040.1 | 7 - 8 | 1763 | 127 | |
| XM_006256933.2 | XP_006256995.1 | 7 - 8 | 1818 | 127 | ||
| XM_006256934.2 | XP_006256996.1 | 7 - 8 | 2094 | 127 | ||
| XM_006256936.2 | XP_006256998.1 | 7 - 8 | 1819 | 127 | ||
| XM_006256937.2 | XP_006256999.1 | 7 - 8 | 1818 | 127 | ||
| XM_006256938.2 | XP_006257000.1 | 7 - 8 | 1818 | 127 | ||
| XM_008773186.1 | XP_008771408.1 | 7 - 8 | 1818 | 127 | ||
| GenBank mRNA | BC166438.1 | - | 7 - 8 | 1763 | 7441789 | 127 |
Set Membership: |
|