Search
Search

Colgalt1| Gene Symbol: Colgalt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106067.2 | NP_001099537.1 | 5 - 6 | 815 | 66 | |
| GenBank mRNA | BC160899.1 | - | 5 - 6 | 815 | 7453718 | 66 |
Set Membership: |
|