Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127590.1 | NP_001121062.1 | 3 - 4 | 243 | 120 | |
XM_006252950.2 | XP_006253012.1 | 3 - 4 | 324 | 120 | ||
XM_006252951.2 | XP_006253013.1 | 3 - 4 | 324 | 120 | ||
XM_008771155.1 | XP_008769377.1 | 3 - 4 | 841 | 120 | ||
GenBank mRNA | BC160887.1 | - | 3 - 4 | 243 | 7387602 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |