Search
Search
| Gene Symbol: Mau2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106077.2 | NP_001099547.2 | 4 - 5 | 472 | 63 | |
| XM_006252877.2 | XP_006252939.1 | 4 - 5 | 508 | 63 | ||
| XM_006252878.2 | XP_006252940.1 | 4 - 5 | 511 | 63 | ||
Set Membership: |
|