Search
Search

Gatad2a| Gene Symbol: Gatad2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013881.2 | NP_001013903.2 | 12 - 13 | 2309 | 70 | |
| XM_006252879.2 | XP_006252941.1 | 11 - 12 | 1989 | 70 | ||
| XM_006252880.2 | XP_006252942.1 | 11 - 12 | 1986 | 70 | ||
| XM_006252881.2 | XP_006252943.1 | 11 - 12 | 1980 | 70 | ||
| XM_006252882.2 | XP_006252944.1 | 11 - 12 | 1977 | 70 | ||
| XM_006252883.2 | XP_006252945.1 | 10 - 11 | 1914 | 70 | ||
| XM_006252884.2 | XP_006252946.1 | 12 - 13 | 2268 | 70 | ||
| XM_006252887.2 | XP_006252949.1 | 11 - 12 | 1801 | 70 | ||
| XM_006252889.2 | XP_006252951.1 | 11 - 12 | 1928 | 70 | ||
| XM_008771089.1 | XP_008769311.1 | 13 - 14 | 2358 | 70 | ||
| XM_008771090.1 | XP_008769312.1 | 12 - 13 | 1988 | 70 | ||
| GenBank mRNA | BC089902.1 | - | 9 - 10 | 1399 | 7383529 | 70 |
Set Membership: |
|