Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01420675_g1
          See other MEF2B GE Assays ›
          Gene Symbol:
          Mef2b
          Gene Name:
          myocyte enhancer factor 2B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 21001204 - 21017445 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01420675_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mef2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017507.1 NP_001017507.1 7 - 8 806 107
          XM_006252927.2 XP_006252989.1 7 - 8 1126 107
          GenBank mRNA BC079361.1 - 7 - 8 806 7109152 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600002128.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mef2b

          Panther Classification:

          Molecular Function -

          MADS box transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          histone deacetylase binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P085936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266736 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136199 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085936 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085936 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P085936 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48157 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14432 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10791078 - -
          Affymetrix Rat Gene 1.0 ST Array 10791071 - -
          Affymetrix Rat Gene 1.0 ST Array 10791077 - -
          Affymetrix Rat Gene 1.0 ST Array 10791079 - -
          Affymetrix Rat Gene 1.0 ST Array 10791072 - -
          Affymetrix Rat Gene 1.0 ST Array 10791075 - -
          Affymetrix Rat Gene 1.0 ST Array 10791076 - -
          Affymetrix Rat Gene 1.0 ST Array 10791074 - -
          Affymetrix Rat Gene 1.0 ST Array 10791073 - -
          Affymetrix Rat Gene 1.1 ST Array 10791070 - -
          Rat Genome 230 2.0 Array 1391797_at - -
          Rat Genome 230 2.0 Array 1391797_PM_at - -
          Affymetrix HT Rat Focus Array 1391797_at - -
          Affymetrix HT Rat Focus Array 1391797_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600016098.rn.1 TC1600002128.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600016100.rn.1 TC1600002128.rn.1 RTA TC
          Rat Expression Set 230 1391797_at - -
          Rat Expression Set 230 1391797_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10791070 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351308 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline