Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01420988_m1
          See other IRAK2 GE Assays ›
          Gene Symbol:
          Irak2
          Gene Name:
          interleukin-1 receptor-associated kinase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 145594575 - 145651066 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01420988_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Irak2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025422.1 NP_001020593.1 12 - 13 1791 128
          XM_008763194.1 XP_008761416.1 12 - 13 1835 128
          GenBank mRNA BC098060.1 - 12 - 13 1791 7458113 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001581.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Irak2

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cytokine-mediated signaling pathway
          toll-like receptor signaling pathway
          protein phosphorylation
          activation of NF-kappaB-inducing kinase activity
          biological_process
          lipopolysaccharide-mediated signaling pathway
          negative regulation of NF-kappaB transcription factor activity
          intracellular signal transduction
          negative regulation of sequence-specific DNA binding transcription factor activity
          protein autophosphorylation
          positive regulation of NF-kappaB transcription factor activity
          interleukin-1-mediated signaling pathway
          response to interleukin-1
          molecular_function
          protein serine/threonine kinase activity
          ATP binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P541884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662122 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65943 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41166 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100589 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77607 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166480 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6026 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52214 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47821 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114239 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P541884 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P541884 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P541884 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P161617 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102113 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103906 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97433 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50616 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62652 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10857903 - -
          Affymetrix Rat Gene 1.0 ST Array 10857892 - -
          Affymetrix Rat Gene 1.0 ST Array 10857901 - -
          Affymetrix Rat Gene 1.0 ST Array 10857896 - -
          Affymetrix Rat Gene 1.0 ST Array 10857899 - -
          Affymetrix Rat Gene 1.0 ST Array 10857904 - -
          Affymetrix Rat Gene 1.0 ST Array 10857898 - -
          Affymetrix Rat Gene 1.0 ST Array 10857895 - -
          Affymetrix Rat Gene 1.0 ST Array 10857900 - -
          Affymetrix Rat Gene 1.0 ST Array 10857893 - -
          Affymetrix Rat Gene 1.0 ST Array 10857902 - -
          Affymetrix Rat Gene 1.0 ST Array 10857906 - -
          Affymetrix Rat Gene 1.0 ST Array 10857905 - -
          Affymetrix Rat Gene 1.0 ST Array 10857897 - -
          Affymetrix Rat Gene 1.0 ST Array 10857907 - -
          Affymetrix Rat Gene 1.0 ST Array 10857894 - -
          Affymetrix Rat Gene 1.1 ST Array 10857891 - -
          Rat Genome 230 2.0 Array 1383474_at - -
          Rat Genome 230 2.0 Array 1383474_PM_at - -
          Affymetrix HT Rat Focus Array 1383474_PM_at - -
          Affymetrix HT Rat Focus Array 1383474_at - -
          Affymetrix Rat Clariom S Assay PSR0400011272.rn.1 TC0400001581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011273.rn.1 TC0400001581.rn.1 RTA TC
          Rat Expression Set 230 1383474_at - -
          Rat Expression Set 230 1383474_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10857891 - -
          Rat Genome U34 Set rc_AI145328_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21382194 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373683 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline