Search
Search
| Gene Symbol: Prok2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037541.1 | NP_001032630.1 | 1 - 2 | 90 | 98 | |
| NM_138852.1 | NP_620207.1 | 1 - 2 | 90 | 98 | ||
| GenBank mRNA | AY089984.1 | - | 1 - 2 | 90 | 98 | |
| AY348322.1 | - | 1 - 2 | 90 | 98 | ||
Set Membership: |
|