Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01421370_m1
          See other IMMT GE Assays ›
          Gene Symbol:
          Immt
          Gene Name:
          inner membrane protein, mitochondrial
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 99753452 - 99789921 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01421370_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Immt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034928.1 NP_001030100.1 13 - 14 1599 113
          XM_008762981.1 XP_008761203.1 14 - 15 1829 113
          XM_008762982.1 XP_008761204.1 13 - 14 1814 113
          XM_008762983.1 XP_008761205.1 14 - 15 1795 113
          XM_008762984.1 XP_008761206.1 14 - 15 1792 113
          XM_008762985.1 XP_008761207.1 13 - 14 1780 113
          XM_008762986.1 XP_008761208.1 13 - 14 1731 113
          XM_008762987.1 XP_008761209.1 13 - 14 1697 113
          XM_008762988.1 XP_008761210.1 13 - 14 1694 113
          XM_008762989.1 XP_008761211.1 12 - 13 1682 113
          XM_008762990.1 XP_008761212.1 12 - 13 1604 113
          GenBank mRNA BC105841.1 - 13 - 14 1599 7454026 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001110.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Immt

          Gene Ontology Categories:

          Function(s) Process(es)

          response to cold
          mitochondrial calcium ion homeostasis
          molecular_function
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P150610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11673 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110348 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135171 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17125 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33948 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P150610 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P150610 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P150610 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1049236 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88520 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103890 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21726 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34400 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10856315 - -
          Affymetrix Rat Gene 1.0 ST Array 10856310 - -
          Affymetrix Rat Gene 1.0 ST Array 10856307 - -
          Affymetrix Rat Gene 1.0 ST Array 10856313 - -
          Affymetrix Rat Gene 1.0 ST Array 10856318 - -
          Affymetrix Rat Gene 1.0 ST Array 10856314 - -
          Affymetrix Rat Gene 1.0 ST Array 10856311 - -
          Affymetrix Rat Gene 1.0 ST Array 10856319 - -
          Affymetrix Rat Gene 1.0 ST Array 10856305 - -
          Affymetrix Rat Gene 1.0 ST Array 10856317 - -
          Affymetrix Rat Gene 1.0 ST Array 10856306 - -
          Affymetrix Rat Gene 1.0 ST Array 10856304 - -
          Affymetrix Rat Gene 1.0 ST Array 10856303 - -
          Affymetrix Rat Gene 1.0 ST Array 10856316 - -
          Affymetrix Rat Gene 1.0 ST Array 10856312 - -
          Affymetrix Rat Gene 1.0 ST Array 10856308 - -
          Affymetrix Rat Gene 1.1 ST Array 10856302 - -
          Rat Genome 230 2.0 Array 1372080_PM_at - -
          Rat Genome 230 2.0 Array 1375597_PM_at - -
          Rat Genome 230 2.0 Array 1372080_at - -
          Rat Genome 230 2.0 Array 1385423_at - -
          Rat Genome 230 2.0 Array 1380154_at - -
          Rat Genome 230 2.0 Array 1375597_at - -
          Rat Genome 230 2.0 Array 1380154_PM_at - -
          Rat Genome 230 2.0 Array 1385423_PM_at - -
          Affymetrix HT Rat Focus Array 1380154_PM_at - -
          Affymetrix HT Rat Focus Array 1380154_at - -
          Affymetrix HT Rat Focus Array 1385423_PM_at - -
          Affymetrix HT Rat Focus Array 1372080_at - -
          Affymetrix HT Rat Focus Array 1385423_at - -
          Affymetrix HT Rat Focus Array 1372080_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400007875.rn.1 TC0400001110.rn.1 RTA TC
          Rat Expression Set 230 1380154_PM_at - -
          Rat Expression Set 230 1380154_at - -
          Rat Expression Set 230 1372080_PM_at - -
          Rat Expression Set 230 1372080_at - -
          Rat Expression Set 230 1385423_at - -
          Rat Expression Set 230 1385423_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10856302 - -
          Rat Genome U34 Set rc_AI639260_at - -
          Rat Genome U34 Set rc_AI176477_at - -
          Rat Genome U34 Set rc_AI231567_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22212002 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369795 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline