Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01421410_m1
          See other RAD18 GE Assays ›
          Gene Symbol:
          Rad18
          Gene Name:
          RAD18 E3 ubiquitin protein ligase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 144537329 - 144620613 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01421410_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rad18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077673.1 NP_001071141.1 4 - 5 319 94
          XM_006237036.1 XP_006237098.1 4 - 5 348 94
          XM_006237037.1 XP_006237099.1 4 - 5 348 94
          GenBank mRNA BC127510.1 - 4 - 5 319 8366167 94
          EF395817.1 - 289 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003516.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          cellular response to DNA damage stimulus
          spermatogenesis
          biological_process
          response to UV
          negative regulation of DNA recombination
          Y-form DNA binding
          molecular_function
          zinc ion binding
          polyubiquitin binding
          ubiquitin protein ligase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P23316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828517 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97922 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126473 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72655 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110476 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65563 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61603 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133390 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142198 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175848 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P23316 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P23316 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P23316 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P23316 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70029 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70707 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10864573 - -
          Affymetrix Rat Gene 1.0 ST Array 10864586 - -
          Affymetrix Rat Gene 1.0 ST Array 10864582 - -
          Affymetrix Rat Gene 1.0 ST Array 10864574 - -
          Affymetrix Rat Gene 1.0 ST Array 10864581 - -
          Affymetrix Rat Gene 1.0 ST Array 10864579 - -
          Affymetrix Rat Gene 1.0 ST Array 10864587 - -
          Affymetrix Rat Gene 1.0 ST Array 10864575 - -
          Affymetrix Rat Gene 1.0 ST Array 10864585 - -
          Affymetrix Rat Gene 1.0 ST Array 10864580 - -
          Affymetrix Rat Gene 1.0 ST Array 10864577 - -
          Affymetrix Rat Gene 1.0 ST Array 10864583 - -
          Affymetrix Rat Gene 1.0 ST Array 10864578 - -
          Affymetrix Rat Gene 1.0 ST Array 10864584 - -
          Affymetrix Rat Gene 1.1 ST Array 10864572 - -
          Rat Genome 230 2.0 Array 1392449_PM_at - -
          Rat Genome 230 2.0 Array 1392762_PM_at - -
          Rat Genome 230 2.0 Array 1392449_at - -
          Rat Genome 230 2.0 Array 1392762_at - -
          Affymetrix HT Rat Focus Array 1392449_at - -
          Affymetrix HT Rat Focus Array 1392449_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400024680.rn.1 TC0400003516.rn.1 RTA TC
          Rat Expression Set 230 1392762_PM_at - -
          Rat Expression Set 230 1392449_at - -
          Rat Expression Set 230 1392449_PM_at - -
          Rat Expression Set 230 1392762_at - -
          Affymetrix Rat Gene 1.0 ST Array 10864572 - -
          Rat Genome U34 Set rc_AA800878_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22011571 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350574 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline