Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01421528_m1
          See other SLC35F2 GE Assays ›
          Gene Symbol:
          Slc35f2
          Gene Name:
          solute carrier family 35, member F2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 58347673 - 58391462 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01421528_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc35f2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106822.1 NP_001100292.1 6 - 7 1028 79
          XM_006243069.2 XP_006243131.1 7 - 8 1272 79
          GenBank mRNA BC091334.1 - 6 - 7 931 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000728.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P000136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937512 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14998 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154503 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114314 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123346 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111401 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63524 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000136 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000136 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P000136 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P250970 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63396 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10910044 - -
          Affymetrix Rat Gene 1.0 ST Array 10910039 - -
          Affymetrix Rat Gene 1.0 ST Array 10910041 - -
          Affymetrix Rat Gene 1.0 ST Array 10910040 - -
          Affymetrix Rat Gene 1.0 ST Array 10910045 - -
          Affymetrix Rat Gene 1.0 ST Array 10910043 - -
          Affymetrix Rat Gene 1.0 ST Array 10910046 - -
          Affymetrix Rat Gene 1.1 ST Array 10910038 - -
          Rat Genome 230 2.0 Array 1398660_at - -
          Rat Genome 230 2.0 Array 1398660_PM_at - -
          Affymetrix HT Rat Focus Array 1398660_PM_at - -
          Affymetrix HT Rat Focus Array 1398660_at - -
          Affymetrix Rat Clariom S Assay PSR0800004803.rn.1 TC0800000728.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800004805.rn.1 TC0800000728.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800004810.rn.1 TC0800000728.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800004804.rn.1 TC0800000728.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800004808.rn.1 TC0800000728.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800004811.rn.1 TC0800000728.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800004812.rn.1 TC0800000728.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800004806.rn.1 TC0800000728.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800004809.rn.1 TC0800000728.rn.1 RTA TC
          Rat Expression Set 230 1398660_PM_at - -
          Rat Expression Set 230 1398660_at - -
          Affymetrix Rat Gene 1.0 ST Array 10910038 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20955554 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359020 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline