Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01421631_m1
          See other SIDT2 GE Assays ›
          Gene Symbol:
          Sidt2
          Gene Name:
          SID1 transmembrane family, member 2
          Gene Aliases:
          RGD1308311
          Chromosome Location:
          Chr.8: 50230335 - 50247187 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01421631_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sidt2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108142.1 NP_001101612.1 1 - 2 996 84
          XM_006242941.2 XP_006243003.1 1 - 2 1527 84
          XM_006242942.2 XP_006243004.1 1 - 2 1527 84
          XM_006242943.2 XP_006243005.1 1 - 2 1526 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002450.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          type B pancreatic cell development
          response to glucose
          dsRNA transport
          glucose homeostasis
          type B pancreatic cell proliferation
          regulation of insulin secretion involved in cellular response to glucose stimulus
          RNA transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P100938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218011 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124967 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26578 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55369 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164437 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15524 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1785 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100938 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100938 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P100938 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P121887 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47143 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35496 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10917057 - -
          Affymetrix Rat Gene 1.0 ST Array 10917048 - -
          Affymetrix Rat Gene 1.0 ST Array 10917063 - -
          Affymetrix Rat Gene 1.0 ST Array 10917041 - -
          Affymetrix Rat Gene 1.0 ST Array 10917067 - -
          Affymetrix Rat Gene 1.0 ST Array 10917042 - -
          Affymetrix Rat Gene 1.0 ST Array 10917046 - -
          Affymetrix Rat Gene 1.0 ST Array 10917064 - -
          Affymetrix Rat Gene 1.0 ST Array 10917047 - -
          Affymetrix Rat Gene 1.0 ST Array 10917055 - -
          Affymetrix Rat Gene 1.0 ST Array 10917059 - -
          Affymetrix Rat Gene 1.0 ST Array 10917054 - -
          Affymetrix Rat Gene 1.0 ST Array 10917066 - -
          Affymetrix Rat Gene 1.0 ST Array 10917061 - -
          Affymetrix Rat Gene 1.0 ST Array 10917051 - -
          Affymetrix Rat Gene 1.0 ST Array 10917044 - -
          Affymetrix Rat Gene 1.0 ST Array 10917056 - -
          Affymetrix Rat Gene 1.0 ST Array 10917050 - -
          Affymetrix Rat Gene 1.0 ST Array 10917049 - -
          Affymetrix Rat Gene 1.0 ST Array 10917045 - -
          Affymetrix Rat Gene 1.0 ST Array 10917065 - -
          Affymetrix Rat Gene 1.0 ST Array 10917052 - -
          Affymetrix Rat Gene 1.0 ST Array 10917058 - -
          Affymetrix Rat Gene 1.0 ST Array 10917068 - -
          Affymetrix Rat Gene 1.0 ST Array 10917043 - -
          Affymetrix Rat Gene 1.0 ST Array 10917062 - -
          Affymetrix Rat Gene 1.0 ST Array 10917053 - -
          Affymetrix Rat Gene 1.0 ST Array 10917060 - -
          Affymetrix Rat Gene 1.1 ST Array 10917040 - -
          Rat Genome 230 2.0 Array 1388828_PM_at - -
          Rat Genome 230 2.0 Array 1388828_at - -
          Affymetrix HT Rat Focus Array 1388828_PM_at - -
          Affymetrix HT Rat Focus Array 1388828_at - -
          Affymetrix Rat Clariom S Assay PSR0800019737.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019711.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019728.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019745.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019746.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019720.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019733.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019721.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019717.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019708.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019723.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019730.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019712.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019707.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019744.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019732.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019722.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019714.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019726.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019740.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019738.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019750.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019752.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019751.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019716.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019739.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019747.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019710.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019743.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019741.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019748.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019709.rn.1 TC0800002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019724.rn.1 TC0800002450.rn.1 RTA TC
          Rat Expression Set 230 1388828_PM_at - -
          Rat Expression Set 230 1388828_at - -
          Affymetrix Rat Gene 1.0 ST Array 10917040 - -
          Rat Genome U34 Set rc_AA956733_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369730 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline