Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01421873_g1
          See other HMBS GE Assays ›
          Gene Symbol:
          Hmbs
          Gene Name:
          hydroxymethylbilane synthase
          Gene Aliases:
          PBG-D, PBGD, URO-S, hemC
          Chromosome Location:
          Chr.8: 48667278 - 48674670 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01421873_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hmbs
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013168.2 NP_037300.2 12 - 13 954 76
          XM_006242896.1 XP_006242958.1 12 - 13 1040 76
          XM_006242897.1 XP_006242959.1 11 - 12 962 76
          XM_008766148.1 XP_008764370.1 11 - 12 1124 76
          XM_008766149.1 XP_008764371.1 11 - 12 1061 76
          GenBank mRNA BC088162.1 - 12 - 13 954 7375228 76
          FQ226062.1 - 12 - 13 877 76
          FQ230962.1 - 6 - 7 439 76
          FQ231091.1 - 12 - 13 939 76
          FQ234793.1 - 4 - 5 326 76
          FQ234870.1 - 10 - 11 1110 76
          X06827.1 - 12 - 13 829 76
          Y12006.1 - 12 - 13 824 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002404.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Endogenous Controls Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hmbs

          Panther Classification:

          Molecular Function -

          deaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          protoporphyrinogen IX biosynthetic process
          response to hormone
          response to carbohydrate
          response to metal ion
          response to zinc ion
          response to lead ion
          response to organic cyclic compound
          peptidyl-pyrromethane cofactor linkage
          organ regeneration
          response to nutrient levels
          response to cobalt ion
          response to estradiol
          tetrapyrrole biosynthetic process
          response to vitamin
          response to drug
          response to amino acid
          astrocyte differentiation
          response to methylmercury
          cellular response to antibiotic
          cellular response to arsenic-containing substance
          cellular response to lead ion
          cellular response to cytokine stimulus
          cellular response to amine stimulus
          cellular response to dexamethasone stimulus
          hydroxymethylbilane synthase activity
          uroporphyrinogen-III synthase activity
          protein binding
          carboxylic acid binding
          amine binding
          coenzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P421363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192297 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P421363 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P421363 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P421363 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P421363 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P262289 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700178371 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58092 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62275 - -
          Agilent Rat cDNA Microarray (G4105A) 700178371 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10916759 - -
          Affymetrix Rat Gene 1.0 ST Array 10916762 - -
          Affymetrix Rat Gene 1.0 ST Array 10916758 - -
          Affymetrix Rat Gene 1.0 ST Array 10916756 - -
          Affymetrix Rat Gene 1.0 ST Array 10916767 - -
          Affymetrix Rat Gene 1.0 ST Array 10916765 - -
          Affymetrix Rat Gene 1.0 ST Array 10916761 - -
          Affymetrix Rat Gene 1.0 ST Array 10916763 - -
          Affymetrix Rat Gene 1.0 ST Array 10916754 - -
          Affymetrix Rat Gene 1.0 ST Array 10916760 - -
          Affymetrix Rat Gene 1.0 ST Array 10916757 - -
          Affymetrix Rat Gene 1.0 ST Array 10916764 - -
          Affymetrix Rat Gene 1.0 ST Array 10916755 - -
          Affymetrix Rat Gene 1.0 ST Array 10916766 - -
          Affymetrix Rat Gene 1.1 ST Array 10916753 - -
          Rat Genome 230 2.0 Array 1386983_at - -
          Rat Genome 230 2.0 Array 1386983_PM_at - -
          Affymetrix HT Rat Focus Array 1386983_PM_at - -
          Affymetrix HT Rat Focus Array 1386983_at - -
          Affymetrix Rat Clariom S Assay PSR0800019163.rn.1 TC0800002404.rn.1 RTA TC
          Rat Expression Set 230 1386983_at - -
          Rat Expression Set 230 1386983_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10916753 - -
          Rat Genome U34 Set X06827_at - -
          Rat Genome U34 Set X06827_g_at - -
          Rat Genome U34 Set X06827_at - -
          Rat Genome U34 Set X06827_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21552446 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353365 - -
          RatRef-12 Expression BeadChip ILMN_66660 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline