Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01422019_m1
          See other SCN3B GE Assays ›
          Gene Symbol:
          Scn3b
          Gene Name:
          sodium channel, voltage-gated, type III, beta subunit
          Gene Aliases:
          Scnb3
          Chromosome Location:
          Chr.8: 44136414 - 44159011 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01422019_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scn3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139097.3 NP_620797.1 3 - 4 597 107
          XM_006243102.1 XP_006243164.1 3 - 4 679 107
          XM_006243103.1 XP_006243165.1 3 - 4 679 107
          XM_008766114.1 XP_008764336.1 3 - 4 679 107
          XM_008766115.1 XP_008764337.1 2 - 3 246 107
          GenBank mRNA AF378093.1 - 3 - 4 296 107
          AJ243395.2 - 3 - 4 582 107
          BC070899.1 - 3 - 4 597 7100476 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000500.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scn3b

          Panther Classification:

          Molecular Function -

          ion channel transporter voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          sodium ion transport
          nervous system development
          positive regulation of heart rate
          positive regulation of sodium ion transport
          sensory perception of pain
          regulation of ion transmembrane transport
          sodium ion transmembrane transport
          membrane depolarization
          cardiac muscle contraction
          regulation of atrial cardiac muscle cell membrane depolarization
          regulation of ventricular cardiac muscle cell membrane depolarization
          cardiac conduction
          protein localization to plasma membrane
          cardiac muscle cell action potential involved in contraction
          ventricular cardiac muscle cell action potential
          membrane depolarization during action potential
          membrane depolarization during cardiac muscle cell action potential
          atrial cardiac muscle cell action potential
          SA node cell to atrial cardiac muscle cell communication
          regulation of heart rate by cardiac conduction
          regulation of sodium ion transmembrane transporter activity
          molecular_function
          sodium channel regulator activity
          ion channel binding
          voltage-gated sodium channel activity involved in cardiac muscle cell action potential

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P779282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371146 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97799 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44914 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2177 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P779282 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P779282 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P779282 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P779282 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702034207 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14754 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14636 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33900 - -
          Agilent Rat cDNA Microarray (G4105A) 702034207 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10909336 - -
          Affymetrix Rat Gene 1.0 ST Array 10909337 - -
          Affymetrix Rat Gene 1.0 ST Array 10909330 - -
          Affymetrix Rat Gene 1.0 ST Array 10909333 - -
          Affymetrix Rat Gene 1.0 ST Array 10909332 - -
          Affymetrix Rat Gene 1.0 ST Array 10909335 - -
          Affymetrix Rat Gene 1.0 ST Array 10909334 - -
          Affymetrix Rat Gene 1.0 ST Array 10909331 - -
          Affymetrix Rat Gene 1.0 ST Array 10909329 - -
          Affymetrix Rat Gene 1.1 ST Array 10909328 - -
          Rat Genome 230 2.0 Array 1383435_at - -
          Rat Genome 230 2.0 Array 1383435_PM_at - -
          Rat Genome 230 2.0 Array 1370850_at - -
          Rat Genome 230 2.0 Array 1370850_PM_at - -
          Affymetrix HT Rat Focus Array 1370850_PM_at - -
          Affymetrix HT Rat Focus Array 1383435_at - -
          Affymetrix HT Rat Focus Array 1370850_at - -
          Affymetrix HT Rat Focus Array 1383435_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800003095.rn.1 TC0800000500.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800003094.rn.1 TC0800000500.rn.1 RTA TC
          Rat Expression Set 230 1370850_at - -
          Rat Expression Set 230 1370850_PM_at - -
          Rat Expression Set 230 1383435_at - -
          Rat Expression Set 230 1383435_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10909328 - -
          Rat Genome U34 Set rc_AA946203_at - -
          Rat Genome U34 Set rc_AA963067_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350770 - -
          RatRef-12 Expression BeadChip ILMN_67120 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0