Search
Search
| Gene Symbol: Treh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136141.1 | NP_001129613.1 | 9 - 10 | 920 | 66 | |
| XM_006242956.2 | XP_006243018.1 | 8 - 9 | 792 | 66 | ||
| GenBank mRNA | AF038043.1 | - | 8 - 9 | 841 | 66 | |
Set Membership: |
|