Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01422317_m1
          See other SNX33 GE Assays ›
          Gene Symbol:
          Snx33
          Gene Name:
          sorting nexin 33
          Gene Aliases:
          Sh3px3
          Chromosome Location:
          Chr.8: 61584656 - 61596056 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01422317_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Snx33
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127488.1 NP_001120960.1 1 - 2 2380 80
          XM_006243123.2 XP_006243185.1 2 - 3 3209 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002617.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          intracellular protein transport
          endocytosis
          endosome organization
          mitotic nuclear division
          vesicle organization
          endosomal transport
          protein import
          cleavage furrow formation
          macropinocytosis
          negative regulation of endocytosis
          positive regulation of membrane protein ectodomain proteolysis
          membrane tubulation
          negative regulation of protein localization to cell surface
          positive regulation of protein localization to cell surface
          phosphatidylinositol binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P403396 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P683816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848003 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9354 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P683816 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P403396 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P403396 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P683816 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P683816 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P403396 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P403396 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P683816 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10917724 - -
          Affymetrix Rat Gene 1.0 ST Array 10917723 - -
          Affymetrix Rat Gene 1.1 ST Array 10917722 - -
          Rat Genome 230 2.0 Array 1373239_PM_at - -
          Rat Genome 230 2.0 Array 1373239_at - -
          Affymetrix HT Rat Focus Array 1373239_PM_at - -
          Affymetrix HT Rat Focus Array 1373239_at - -
          Affymetrix Rat Clariom S Assay PSR0800021177.rn.1 TC0800002617.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021178.rn.1 TC0800002617.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021180.rn.1 TC0800002617.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021179.rn.1 TC0800002617.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021181.rn.1 TC0800002617.rn.1 RTA TC
          Rat Expression Set 230 1373239_at - -
          Rat Expression Set 230 1373239_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10917722 - -
          Rat Genome U34 Set rc_AI071703_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22247528 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351211 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline