Search
Search
| Gene Symbol: Arid3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109001.1 | NP_001102471.1 | 6 - 7 | 1492 | 64 | |
| XM_006243171.2 | XP_006243233.1 | 7 - 8 | 1589 | 64 | ||
| XM_008766323.1 | XP_008764545.1 | 7 - 8 | 3736 | 64 | ||
Set Membership: |
|