Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01422596_m1
          See other SERPINB13 GE Assays ›
          Gene Symbol:
          Serpinb13
          Gene Name:
          serpin peptidase inhibitor, clade B (ovalbumin), member 13
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 27032048 - 27062620 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01422596_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpinb13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107168.1 NP_001100638.1 7 - 8 827 108
          XM_008769454.1 XP_008767676.1 7 - 8 827 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000089.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endopeptidase activity
          negative regulation of keratinocyte apoptotic process
          protease binding
          serine-type endopeptidase inhibitor activity
          cysteine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383499 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96361 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86957 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89386 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102254 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047842 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047842 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047842 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P238676 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55677 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10763348 - -
          Affymetrix Rat Gene 1.0 ST Array 10763347 - -
          Affymetrix Rat Gene 1.0 ST Array 10763344 - -
          Affymetrix Rat Gene 1.0 ST Array 10763350 - -
          Affymetrix Rat Gene 1.0 ST Array 10763345 - -
          Affymetrix Rat Gene 1.0 ST Array 10763346 - -
          Affymetrix Rat Gene 1.0 ST Array 10763349 - -
          Affymetrix Rat Gene 1.1 ST Array 10763342 - -
          Affymetrix Rat Clariom S Assay PSR1300000443.rn.1 TC1300000089.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300000448.rn.1 TC1300000089.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10763342 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22078840 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350373 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline