Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01423020_m1
          See other SLC9A8 GE Assays ›
          Gene Symbol:
          Slc9a8
          Gene Name:
          solute carrier family 9, subfamily A (NHE8, cation proton antiporter 8), member 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 164173059 - 164224747 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01423020_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc9a8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025281.1 NP_001020452.1 14 - 15 1504 71
          XM_006235645.2 XP_006235707.1 13 - 14 2658 71
          XM_006235646.2 XP_006235708.1 14 - 15 2820 71
          GenBank mRNA AY496958.1 - 14 - 15 1504 71
          FQ210940.1 - 12 - 13 1260 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002364.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc9a8

          Panther Classification:

          Molecular Function -

          transporter secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          potassium ion transport
          sodium ion transport
          regulation of pH
          sodium ion transmembrane transport
          regulation of intracellular pH
          potassium ion transmembrane transport
          hydrogen ion transmembrane transport
          sodium:proton antiporter activity
          potassium:proton antiporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P269808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782381 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116173 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66802 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4513 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P269808 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P269808 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P269808 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P269808 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15174 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40211 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36414 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37768 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25220 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10842452 - -
          Affymetrix Rat Gene 1.0 ST Array 10842441 - -
          Affymetrix Rat Gene 1.0 ST Array 10842448 - -
          Affymetrix Rat Gene 1.0 ST Array 10842450 - -
          Affymetrix Rat Gene 1.0 ST Array 10842456 - -
          Affymetrix Rat Gene 1.0 ST Array 10842442 - -
          Affymetrix Rat Gene 1.0 ST Array 10842447 - -
          Affymetrix Rat Gene 1.0 ST Array 10842443 - -
          Affymetrix Rat Gene 1.0 ST Array 10842451 - -
          Affymetrix Rat Gene 1.0 ST Array 10842453 - -
          Affymetrix Rat Gene 1.0 ST Array 10842445 - -
          Affymetrix Rat Gene 1.0 ST Array 10842454 - -
          Affymetrix Rat Gene 1.0 ST Array 10842446 - -
          Affymetrix Rat Gene 1.0 ST Array 10842455 - -
          Affymetrix Rat Gene 1.0 ST Array 10842444 - -
          Affymetrix Rat Gene 1.0 ST Array 10842449 - -
          Affymetrix Rat Gene 1.1 ST Array 10842440 - -
          Affymetrix Rat Clariom S Assay PSR0300017708.rn.1 TC0300002364.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10842440 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22168497 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360784 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline