Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01423040_m1
          See other EYA2 GE Assays ›
          Gene Symbol:
          Eya2
          Gene Name:
          EYA transcriptional coactivator and phosphatase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 162285279 - 162470642 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01423040_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eya2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130427.1 NP_569111.1 9 - 10 958 129
          XM_008762456.1 XP_008760678.1 10 - 11 1194 129
          GenBank mRNA AY366465.1 - 9 - 10 958 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002324.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eya2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          multicellular organism development
          striated muscle tissue development
          histone dephosphorylation
          peptidyl-tyrosine dephosphorylation
          extrinsic apoptotic signaling pathway in absence of ligand
          mitochondrial outer membrane permeabilization
          magnesium ion binding
          protein tyrosine phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P069664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319706 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83549 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94962 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96541 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174841 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136432 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24890 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141452 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107257 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141229 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75615 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156581 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133898 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96661 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108265 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167981 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069664 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069664 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P069664 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P262263 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42455 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28431 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27210 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20894 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21392 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16423 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49464 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4165 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76574 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10842301 - -
          Affymetrix Rat Gene 1.0 ST Array 10842302 - -
          Affymetrix Rat Gene 1.0 ST Array 10842309 - -
          Affymetrix Rat Gene 1.0 ST Array 10842306 - -
          Affymetrix Rat Gene 1.0 ST Array 10842312 - -
          Affymetrix Rat Gene 1.0 ST Array 10842304 - -
          Affymetrix Rat Gene 1.0 ST Array 10842305 - -
          Affymetrix Rat Gene 1.0 ST Array 10842300 - -
          Affymetrix Rat Gene 1.0 ST Array 10842303 - -
          Affymetrix Rat Gene 1.0 ST Array 10842314 - -
          Affymetrix Rat Gene 1.0 ST Array 10842313 - -
          Affymetrix Rat Gene 1.0 ST Array 10842308 - -
          Affymetrix Rat Gene 1.0 ST Array 10842311 - -
          Affymetrix Rat Gene 1.0 ST Array 10842307 - -
          Affymetrix Rat Gene 1.0 ST Array 10842310 - -
          Affymetrix Rat Gene 1.0 ST Array 10842299 - -
          Affymetrix Rat Gene 1.1 ST Array 10842298 - -
          Rat Genome 230 2.0 Array 1386666_at - -
          Rat Genome 230 2.0 Array 1386666_PM_at - -
          Rat Genome 230 2.0 Array 1383610_at - -
          Rat Genome 230 2.0 Array 1383610_PM_at - -
          Affymetrix HT Rat Focus Array 1386666_at - -
          Affymetrix HT Rat Focus Array 1386666_PM_at - -
          Affymetrix HT Rat Focus Array 1383610_at - -
          Affymetrix HT Rat Focus Array 1383610_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300017390.rn.1 TC0300002324.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300017394.rn.1 TC0300002324.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300017392.rn.1 TC0300002324.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300017393.rn.1 TC0300002324.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300017389.rn.1 TC0300002324.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300017388.rn.1 TC0300002324.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300017391.rn.1 TC0300002324.rn.1 RTA TC
          Rat Expression Set 230 1386666_at - -
          Rat Expression Set 230 1383610_at - -
          Rat Expression Set 230 1386666_PM_at - -
          Rat Expression Set 230 1383610_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10842298 - -
          Rat Genome U34 Set rc_AI104875_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360125 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline