Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01423088_m1
          See other SERINC3 GE Assays ›
          Gene Symbol:
          Serinc3
          Gene Name:
          serine incorporator 3
          Gene Aliases:
          Tde1
          Chromosome Location:
          Chr.3: 160018643 - 160038078 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01423088_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serinc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008312.1 NP_001008313.1 9 - 10 1371 71
          GenBank mRNA BC085853.1 - 9 - 10 1371 7190015 71
          EU220431.1 - 9 - 10 1317 71
          FQ213438.1 - 1 - 2 232 71
          FQ222836.1 - 3 - 4 454 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004714.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serinc3

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidylserine metabolic process
          sphingolipid metabolic process
          L-serine transport
          positive regulation of transferase activity
          positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          L-serine transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P156922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48055 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87069 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68953 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5227 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P156922 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P156922 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P156922 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P156922 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98338 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91539 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33684 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10851504 - -
          Affymetrix Rat Gene 1.0 ST Array 10851503 - -
          Affymetrix Rat Gene 1.0 ST Array 10851511 - -
          Affymetrix Rat Gene 1.0 ST Array 10851512 - -
          Affymetrix Rat Gene 1.0 ST Array 10851510 - -
          Affymetrix Rat Gene 1.0 ST Array 10851509 - -
          Affymetrix Rat Gene 1.0 ST Array 10851508 - -
          Affymetrix Rat Gene 1.0 ST Array 10851505 - -
          Affymetrix Rat Gene 1.0 ST Array 10851507 - -
          Affymetrix Rat Gene 1.0 ST Array 10851506 - -
          Affymetrix Rat Gene 1.1 ST Array 10851502 - -
          Rat Genome 230 2.0 Array 1398986_PM_at - -
          Rat Genome 230 2.0 Array 1398986_at - -
          Affymetrix HT Rat Focus Array 1398986_PM_at - -
          Affymetrix HT Rat Focus Array 1398986_at - -
          Affymetrix Rat Clariom S Assay PSR0300036109.rn.1 TC0300004714.rn.1 RTA TC
          Rat Expression Set 230 1398986_PM_at - -
          Rat Expression Set 230 1398986_at - -
          Affymetrix Rat Gene 1.0 ST Array 10851502 - -
          Rat Genome U34 Set rc_AA799641_g_at - -
          Rat Genome U34 Set rc_AA799641_at - -
          Rat Genome U34 Set rc_AA945757_at - -
          Rat Genome U34 Set rc_AI104474_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365634 - -
          RatRef-12 Expression BeadChip ILMN_62295 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline