Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01423169_m1
See other NCOA3 GE Assays ›
Gene Symbol:
Ncoa3
Gene Name:
nuclear receptor coactivator 3
Gene Aliases:
Aib1, Tram-1
Chromosome Location:
Chr.3: 162692176 - 162788582 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01423169_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Ncoa3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_006235634.1
XP_006235696.1
10 - 11
1338
67
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Recommended Assay
Rattus norvegicus
Inventoried
Probe spans exons
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
male gonad development
positive regulation of gene expression
histone acetylation
positive regulation of cell growth
cellular response to hormone stimulus
positive regulation of intracellular steroid hormone receptor signaling pathway
positive regulation of intracellular estrogen receptor signaling pathway
multicellular organism growth
negative regulation of apoptotic process
positive regulation of keratinocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
developmental growth
positive regulation of sequence-specific DNA binding transcription factor activity
vagina development
labyrinthine layer morphogenesis
mammary gland branching involved in thelarche
regulation of RNA biosynthetic process
RNA polymerase II regulatory region DNA binding
chromatin binding
transcription coactivator activity
histone acetyltransferase activity
signal transducer activity
ligand-dependent nuclear receptor binding
estrogen receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
protein complex binding
nuclear hormone receptor binding
peroxisome proliferator activated receptor binding
protein dimerization activity
protein N-terminus binding
androgen receptor binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline