Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01423447_g1
          See other SPATA25 GE Assays ›
          Gene Symbol:
          Spata25
          Gene Name:
          spermatogenesis associated 25
          Gene Aliases:
          C20orf165
          Chromosome Location:
          Chr.3: 161292170 - 161294125 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01423447_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spata25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025769.1 NP_001020940.1 1 - 2 103 87
          GenBank mRNA BC097436.1 - 1 - 2 103 7455517 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004759.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spata25

          Gene Ontology Categories:

          Function(s)

          spermatogenesis

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P142992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570439 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32505 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142992 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142992 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P142992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15755 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42650 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40132 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10851666 - -
          Affymetrix Rat Gene 1.0 ST Array 10851665 - -
          Affymetrix Rat Gene 1.1 ST Array 10851664 - -
          Affymetrix Rat Clariom S Assay PSR0300036431.rn.1 TC0300004759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036432.rn.1 TC0300004759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036427.rn.1 TC0300004759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036428.rn.1 TC0300004759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036426.rn.1 TC0300004759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036429.rn.1 TC0300004759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036430.rn.1 TC0300004759.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10851664 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21567354 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373871 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358612 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline