Search
Search
| Gene Symbol: Dbndd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047111.2 | NP_001040576.1 | 2 - 3 | 534 | 144 | |
| XM_006235610.1 | XP_006235672.1 | 3 - 4 | 597 | 144 | ||
| XM_006235612.1 | XP_006235674.1 | 3 - 4 | 353 | 144 | ||
| XM_006235614.2 | XP_006235676.1 | 3 - 4 | 496 | 144 | ||
| XM_006235615.2 | XP_006235677.1 | 3 - 4 | 442 | 144 | ||
| GenBank mRNA | AY113696.1 | - | 2 - 3 | 412 | 144 | |
| BC158561.1 | - | 2 - 3 | 534 | 7387488 | 144 | |
| FQ213987.1 | - | 2 - 3 | 144 | |||
Set Membership: |
|