Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01423813_g1
          See other TNNC2 GE Assays ›
          Gene Symbol:
          Tnnc2
          Gene Name:
          troponin C type 2 (fast)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 161243747 - 161244831 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01423813_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnnc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037351.1 NP_001032428.1 3 - 4 308 71
          GenBank mRNA DQ273679.1 - 3 - 4 308 71
          FQ215069.1 - 372 71
          FQ216531.1 - 372 71
          FQ216869.1 - 372 71
          FQ216875.1 - 402 71
          FQ216940.1 - 364 71
          FQ216956.1 - 385 71
          FQ217047.1 - 372 71
          FQ217054.1 - 372 71
          FQ217106.1 - 401 71
          FQ217137.1 - 372 71
          FQ217169.1 - 372 71
          FQ217172.1 - 372 71
          FQ217195.1 - 372 71
          FQ217236.1 - 383 71
          FQ217299.1 - 372 71
          FQ217378.1 - 372 71
          FQ217706.1 - 372 71
          FQ217788.1 - 372 71
          FQ217840.1 - 372 71
          FQ217842.1 - 372 71
          FQ217896.1 - 371 71
          FQ217902.1 - 372 71
          FQ217974.1 - 401 71
          FQ217984.1 - 372 71
          FQ218010.1 - 372 71
          FQ218048.1 - 402 71
          FQ218061.1 - 372 71
          FQ223543.1 - 372 71
          FQ223572.1 - 372 71
          FQ223583.1 - 372 71
          FQ223607.1 - 372 71
          FQ223623.1 - 372 71
          FQ223636.1 - 372 71
          FQ223690.1 - 372 71
          FQ223692.1 - 372 71
          FQ223711.1 - 372 71
          FQ223713.1 - 372 71
          FQ223719.1 - 372 71
          FQ223773.1 - 384 71
          FQ223816.1 - 393 71
          FQ223825.1 - 372 71
          FQ223832.1 - 372 71
          FQ223835.1 - 402 71
          FQ223976.1 - 372 71
          FQ223986.1 - 372 71
          FQ224079.1 - 372 71
          FQ224128.1 - 372 71
          FQ224223.1 - 372 71
          FQ224258.1 - 372 71
          FQ224340.1 - 372 71
          FQ224378.1 - 372 71
          FQ224387.1 - 372 71
          FQ224450.1 - 372 71
          FQ224462.1 - 372 71
          FQ224599.1 - 372 71
          FQ224648.1 - 372 71
          FQ224749.1 - 372 71
          FQ224755.1 - 401 71
          FQ224795.1 - 372 71
          FQ224811.1 - 372 71
          FQ224822.1 - 372 71
          FQ224827.1 - 369 71
          J00793.1 - 1 - 2 92 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004755.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnnc2

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal muscle contraction
          actin binding
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P029486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849678 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029486 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029486 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P029486 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P778184 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10851652 - -
          Affymetrix Rat Gene 1.0 ST Array 10851654 - -
          Affymetrix Rat Gene 1.0 ST Array 10851651 - -
          Affymetrix Rat Gene 1.0 ST Array 10851655 - -
          Affymetrix Rat Gene 1.0 ST Array 10851653 - -
          Affymetrix Rat Gene 1.1 ST Array 10851650 - -
          Rat Genome 230 2.0 Array 1372195_at - -
          Rat Genome 230 2.0 Array 1372195_PM_at - -
          Affymetrix HT Rat Focus Array 1372195_PM_at - -
          Affymetrix HT Rat Focus Array 1372195_at - -
          Affymetrix Rat Clariom S Assay PSR0300036393.rn.1 TC0300004755.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036392.rn.1 TC0300004755.rn.1 RTA TC
          Rat Expression Set 230 1372195_at - -
          Rat Expression Set 230 1372195_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10851650 - -
          Rat Genome U34 Set rc_AI639532_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21332185 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373449 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline