Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01423853_m1
          See other TOMM34 GE Assays ›
          Gene Symbol:
          Tomm34
          Gene Name:
          translocase of outer mitochondrial membrane 34
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 160448294 - 160465143 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01423853_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tomm34
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001044244.2 NP_001037709.1 5 - 6 720 63
          XM_006235585.2 XP_006235647.1 5 - 6 995 63
          GenBank mRNA BC105768.1 - 5 - 6 721 7930355 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004725.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to mitochondrion
          heat shock protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P141717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703607 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142275 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142827 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141717 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141717 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P141717 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P323843 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70359 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18207 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43639 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10851535 - -
          Affymetrix Rat Gene 1.0 ST Array 10851538 - -
          Affymetrix Rat Gene 1.0 ST Array 10851540 - -
          Affymetrix Rat Gene 1.0 ST Array 10851539 - -
          Affymetrix Rat Gene 1.0 ST Array 10851537 - -
          Affymetrix Rat Gene 1.0 ST Array 10851536 - -
          Affymetrix Rat Gene 1.0 ST Array 10851541 - -
          Affymetrix Rat Gene 1.0 ST Array 10851543 - -
          Affymetrix Rat Gene 1.1 ST Array 10851534 - -
          Rat Genome 230 2.0 Array 1378729_PM_at - -
          Rat Genome 230 2.0 Array 1389471_PM_at - -
          Rat Genome 230 2.0 Array 1378729_at - -
          Rat Genome 230 2.0 Array 1386439_at - -
          Rat Genome 230 2.0 Array 1386439_PM_at - -
          Rat Genome 230 2.0 Array 1389471_at - -
          Affymetrix HT Rat Focus Array 1378729_at - -
          Affymetrix HT Rat Focus Array 1378729_PM_at - -
          Affymetrix HT Rat Focus Array 1386439_at - -
          Affymetrix HT Rat Focus Array 1389471_PM_at - -
          Affymetrix HT Rat Focus Array 1389471_at - -
          Affymetrix HT Rat Focus Array 1386439_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300036164.rn.1 TC0300004725.rn.1 RTA TC
          Rat Expression Set 230 1386439_at - -
          Rat Expression Set 230 1389471_at - -
          Rat Expression Set 230 1386439_PM_at - -
          Rat Expression Set 230 1389471_PM_at - -
          Rat Expression Set 230 1378729_at - -
          Rat Expression Set 230 1378729_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10851534 - -
          Rat Genome U34 Set rc_H31323_at - -
          Rat Genome U34 Set rc_AI231571_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20853281 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366369 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline