Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01423997_m1
          See other PREX1 GE Assays ›
          Gene Symbol:
          Prex1
          Gene Name:
          phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 1
          Gene Aliases:
          RGD1306534
          Chromosome Location:
          Chr.3: 163329580 - 163477822 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01423997_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prex1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135718.1 NP_001129190.1 6 - 7 872 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004810.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          T cell differentiation
          neutrophil chemotaxis
          regulation of actin filament polymerization
          intracellular signal transduction
          positive regulation of cell adhesion
          regulation of dendrite development
          reactive oxygen species metabolic process
          positive regulation of substrate adhesion-dependent cell spreading
          molecular_function
          Rho guanyl-nucleotide exchange factor activity
          phospholipid binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P288548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666584 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79330 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154867 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42512 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136928 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173334 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20036 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137192 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6343 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92528 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175020 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5059 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67084 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113027 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178037 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1752 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41465 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P288548 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P288548 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P288548 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P288548 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10851880 - -
          Affymetrix Rat Gene 1.0 ST Array 10851891 - -
          Affymetrix Rat Gene 1.0 ST Array 10851871 - -
          Affymetrix Rat Gene 1.0 ST Array 10851893 - -
          Affymetrix Rat Gene 1.0 ST Array 10851887 - -
          Affymetrix Rat Gene 1.0 ST Array 10851866 - -
          Affymetrix Rat Gene 1.0 ST Array 10851868 - -
          Affymetrix Rat Gene 1.0 ST Array 10851870 - -
          Affymetrix Rat Gene 1.0 ST Array 10851867 - -
          Affymetrix Rat Gene 1.0 ST Array 10851894 - -
          Affymetrix Rat Gene 1.0 ST Array 10851863 - -
          Affymetrix Rat Gene 1.0 ST Array 10851905 - -
          Affymetrix Rat Gene 1.0 ST Array 10851878 - -
          Affymetrix Rat Gene 1.0 ST Array 10851888 - -
          Affymetrix Rat Gene 1.0 ST Array 10851865 - -
          Affymetrix Rat Gene 1.0 ST Array 10851864 - -
          Affymetrix Rat Gene 1.0 ST Array 10851902 - -
          Affymetrix Rat Gene 1.0 ST Array 10851869 - -
          Affymetrix Rat Gene 1.0 ST Array 10851885 - -
          Affymetrix Rat Gene 1.0 ST Array 10851901 - -
          Affymetrix Rat Gene 1.0 ST Array 10851875 - -
          Affymetrix Rat Gene 1.0 ST Array 10851882 - -
          Affymetrix Rat Gene 1.0 ST Array 10851890 - -
          Affymetrix Rat Gene 1.0 ST Array 10851873 - -
          Affymetrix Rat Gene 1.0 ST Array 10851898 - -
          Affymetrix Rat Gene 1.0 ST Array 10851900 - -
          Affymetrix Rat Gene 1.0 ST Array 10851876 - -
          Affymetrix Rat Gene 1.0 ST Array 10851903 - -
          Affymetrix Rat Gene 1.0 ST Array 10851896 - -
          Affymetrix Rat Gene 1.0 ST Array 10851872 - -
          Affymetrix Rat Gene 1.0 ST Array 10851874 - -
          Affymetrix Rat Gene 1.0 ST Array 10851881 - -
          Affymetrix Rat Gene 1.0 ST Array 10851899 - -
          Affymetrix Rat Gene 1.0 ST Array 10851889 - -
          Affymetrix Rat Gene 1.0 ST Array 10851897 - -
          Affymetrix Rat Gene 1.0 ST Array 10851906 - -
          Affymetrix Rat Gene 1.0 ST Array 10851883 - -
          Affymetrix Rat Gene 1.0 ST Array 10851904 - -
          Affymetrix Rat Gene 1.0 ST Array 10851886 - -
          Affymetrix Rat Gene 1.0 ST Array 10851895 - -
          Affymetrix Rat Gene 1.0 ST Array 10851892 - -
          Affymetrix Rat Gene 1.0 ST Array 10851884 - -
          Affymetrix Rat Gene 1.0 ST Array 10851879 - -
          Affymetrix Rat Gene 1.1 ST Array 10851861 - -
          Rat Genome 230 2.0 Array 1372055_at - -
          Rat Genome 230 2.0 Array 1372055_PM_at - -
          Rat Genome 230 2.0 Array 1392436_at - -
          Rat Genome 230 2.0 Array 1392436_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300036926.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036898.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036932.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036911.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036916.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036927.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036910.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036933.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036905.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036925.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036931.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036913.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036899.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036924.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036900.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036903.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036940.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036909.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036908.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036936.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036944.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036921.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036901.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036896.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036912.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036938.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036919.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036923.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036907.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036929.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036934.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036918.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036922.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036904.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036902.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036939.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036928.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036917.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036935.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036937.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036941.rn.1 TC0300004810.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036897.rn.1 TC0300004810.rn.1 RTA TC
          Rat Expression Set 230 1372055_PM_at - -
          Rat Expression Set 230 1372055_at - -
          Rat Expression Set 230 1392436_PM_at - -
          Rat Expression Set 230 1392436_at - -
          Affymetrix Rat Gene 1.0 ST Array 10851861 - -
          Rat Genome U34 Set rc_AI060225_at - -
          Rat Genome U34 Set rc_AI013139_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373562 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline