Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01424687_m1
          See other DNAJA3 GE Assays ›
          Gene Symbol:
          Dnaja3
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily A, member 3
          Gene Aliases:
          Tid-1
          Chromosome Location:
          Chr.10: 11059701 - 11085182 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01424687_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnaja3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001038595.1 NP_001033684.1 9 - 10 1238 75
          NM_001038596.1 NP_001033685.1 9 - 10 1238 75
          XM_006245817.2 XP_006245879.1 9 - 10 1278 75
          GenBank mRNA AF516338.1 - 9 - 10 1238 75
          AY077460.1 - 9 - 10 1238 75
          AY264842.1 - 8 - 9 1085 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004392.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          mitochondrial DNA replication
          protein folding
          negative regulation of protein kinase activity
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          activation-induced cell death of T cells
          mitochondrion organization
          small GTPase mediated signal transduction
          neuromuscular junction development
          cell aging
          negative regulation of cell proliferation
          response to heat
          positive regulation of protein ubiquitination
          negative regulation of NF-kappaB transcription factor activity
          T cell differentiation in thymus
          response to interferon-gamma
          protein refolding
          positive regulation of T cell proliferation
          regulation of apoptotic process
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of programmed cell death
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of catalytic activity
          protein stabilization
          negative regulation of interferon-gamma-mediated signaling pathway
          skeletal muscle acetylcholine-gated channel clustering
          interferon-gamma receptor binding
          ATP binding
          transcription factor binding
          protein kinase binding
          Hsp70 protein binding
          receptor tyrosine kinase binding
          metal ion binding
          NF-kappaB binding
          unfolded protein binding
          chaperone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P024267 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P042991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409839 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54357 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147236 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173162 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16605 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042991 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042991 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16605 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P16605 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P042991 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P208880 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83164 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41773 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13037 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10740692 - -
          Affymetrix Rat Gene 1.0 ST Array 10740689 - -
          Affymetrix Rat Gene 1.0 ST Array 10740691 - -
          Affymetrix Rat Gene 1.0 ST Array 10740690 - -
          Affymetrix Rat Gene 1.0 ST Array 10740694 - -
          Affymetrix Rat Gene 1.0 ST Array 10740697 - -
          Affymetrix Rat Gene 1.0 ST Array 10740695 - -
          Affymetrix Rat Gene 1.0 ST Array 10740699 - -
          Affymetrix Rat Gene 1.0 ST Array 10740698 - -
          Affymetrix Rat Gene 1.0 ST Array 10740693 - -
          Affymetrix Rat Gene 1.0 ST Array 10740696 - -
          Affymetrix Rat Gene 1.0 ST Array 10740700 - -
          Affymetrix Rat Gene 1.1 ST Array 10740688 - -
          Rat Genome 230 2.0 Array 1378392_PM_at - -
          Rat Genome 230 2.0 Array 1378392_at - -
          Rat Genome 230 2.0 Array 1389845_PM_at - -
          Rat Genome 230 2.0 Array 1389845_at - -
          Affymetrix HT Rat Focus Array 1378392_at - -
          Affymetrix HT Rat Focus Array 1378392_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000041140.rn.1 TC1000004392.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041139.rn.1 TC1000004392.rn.1 RTA TC
          Rat Expression Set 230 1378392_at - -
          Rat Expression Set 230 1378392_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10740688 - -
          Rat Genome U34 Set S81433_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21259124 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356684 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline