Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01424884_m1
          See other ANKS3 GE Assays ›
          Gene Symbol:
          Anks3
          Gene Name:
          ankyrin repeat and sterile alpha motif domain containing 3
          Gene Aliases:
          RGD1305833
          Chromosome Location:
          Chr.10: 10808683 - 10829507 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01424884_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Anks3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009676.1 NP_001009676.1 11 - 12 1452 70
          XM_006245788.1 XP_006245850.1 11 - 12 1495 70
          XM_006245789.2 XP_006245851.1 11 - 12 1495 70
          XM_006245790.1 XP_006245852.1 11 - 12 1388 70
          XM_006245791.1 XP_006245853.1 8 - 9 1099 70
          XM_008767482.1 XP_008765704.1 11 - 12 1495 70
          XM_008767483.1 XP_008765705.1 10 - 11 3224 70
          XM_008767484.1 XP_008765706.1 9 - 10 1099 70
          GenBank mRNA BC087062.1 - 11 - 12 1452 7131099 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000128.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Anks3

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P590466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661174 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174432 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50106 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P590466 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P590466 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P590466 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20550 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P590466 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43666 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103211 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10731719 - -
          Affymetrix Rat Gene 1.0 ST Array 10731713 - -
          Affymetrix Rat Gene 1.0 ST Array 10731723 - -
          Affymetrix Rat Gene 1.0 ST Array 10731718 - -
          Affymetrix Rat Gene 1.0 ST Array 10731715 - -
          Affymetrix Rat Gene 1.0 ST Array 10731717 - -
          Affymetrix Rat Gene 1.0 ST Array 10731728 - -
          Affymetrix Rat Gene 1.0 ST Array 10731712 - -
          Affymetrix Rat Gene 1.0 ST Array 10731726 - -
          Affymetrix Rat Gene 1.0 ST Array 10731714 - -
          Affymetrix Rat Gene 1.0 ST Array 10731721 - -
          Affymetrix Rat Gene 1.0 ST Array 10731720 - -
          Affymetrix Rat Gene 1.0 ST Array 10731725 - -
          Affymetrix Rat Gene 1.0 ST Array 10731722 - -
          Affymetrix Rat Gene 1.0 ST Array 10731716 - -
          Affymetrix Rat Gene 1.0 ST Array 10731727 - -
          Affymetrix Rat Gene 1.0 ST Array 10731724 - -
          Affymetrix Rat Gene 1.1 ST Array 10731711 - -
          Rat Genome 230 2.0 Array 1376638_at - -
          Rat Genome 230 2.0 Array 1376638_PM_at - -
          Affymetrix HT Rat Focus Array 1376638_at - -
          Affymetrix HT Rat Focus Array 1376638_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000001152.rn.1 TC1000000128.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000001151.rn.1 TC1000000128.rn.1 RTA TC
          Rat Expression Set 230 1376638_PM_at - -
          Rat Expression Set 230 1376638_at - -
          Affymetrix Rat Gene 1.0 ST Array 10731711 - -
          Rat Genome U34 Set rc_AA925004_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21959485 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371755 - -
          RatRef-12 Expression BeadChip ILMN_49641 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline