Search
Search
| Gene Symbol: Usp39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106597.1 | NP_001100067.1 | 2 - 3 | 363 | 87 | |
| XM_006236659.2 | XP_006236721.1 | 3 - 4 | 1392 | 87 | ||
| GenBank mRNA | BC166516.1 | - | 821 | 7131845 | 87 | |
Set Membership: |
|