Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01425934_m1
          See other SETD5 GE Assays ›
          Gene Symbol:
          Setd5
          Gene Name:
          SET domain containing 5
          Gene Aliases:
          RGD1310433
          Chromosome Location:
          Chr.4: 145017608 - 145095247 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01425934_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Setd5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106614.1 NP_001100084.1 20 - 21 4006 64
          XM_006237012.2 XP_006237074.1 19 - 20 3673 64
          XM_006237013.2 XP_006237075.1 17 - 18 7792 64
          XM_006237014.2 XP_006237076.1 18 - 19 7774 64
          XM_006237015.1 XP_006237077.1 19 - 20 3435 64
          XM_006237016.2 XP_006237078.1 18 - 19 3634 64
          XM_006237017.2 XP_006237079.1 17 - 18 7735 64
          XM_006237018.2 XP_006237080.1 18 - 19 3577 64
          XM_006237020.2 XP_006237082.1 18 - 19 4697 64
          XM_006237021.2 XP_006237083.1 19 - 20 3618 64
          XM_008763184.1 XP_008761406.1 18 - 19 7831 64
          XM_008763185.1 XP_008761407.1 19 - 20 7032 64
          XM_008763186.1 XP_008761408.1 19 - 20 4523 64
          XM_008763187.1 XP_008761409.1 18 - 19 4561 64
          XM_008763188.1 XP_008761410.1 19 - 20 4467 64
          XM_008763189.1 XP_008761411.1 16 - 17 3227 64
          GenBank mRNA FQ223178.1 - 19 - 20 3463 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001563.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P869639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846416 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121693 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19449 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104912 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50879 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15259 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97147 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115823 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P869639 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P869639 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P869639 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18992 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P302417 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P869639 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89860 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10857695 - -
          Affymetrix Rat Gene 1.0 ST Array 10857676 - -
          Affymetrix Rat Gene 1.0 ST Array 10857675 - -
          Affymetrix Rat Gene 1.0 ST Array 10857688 - -
          Affymetrix Rat Gene 1.0 ST Array 10857694 - -
          Affymetrix Rat Gene 1.0 ST Array 10857681 - -
          Affymetrix Rat Gene 1.0 ST Array 10857690 - -
          Affymetrix Rat Gene 1.0 ST Array 10857692 - -
          Affymetrix Rat Gene 1.0 ST Array 10857691 - -
          Affymetrix Rat Gene 1.0 ST Array 10857684 - -
          Affymetrix Rat Gene 1.0 ST Array 10857677 - -
          Affymetrix Rat Gene 1.0 ST Array 10857682 - -
          Affymetrix Rat Gene 1.0 ST Array 10857679 - -
          Affymetrix Rat Gene 1.0 ST Array 10857674 - -
          Affymetrix Rat Gene 1.0 ST Array 10857693 - -
          Affymetrix Rat Gene 1.0 ST Array 10857686 - -
          Affymetrix Rat Gene 1.0 ST Array 10857680 - -
          Affymetrix Rat Gene 1.0 ST Array 10857685 - -
          Affymetrix Rat Gene 1.0 ST Array 10857687 - -
          Affymetrix Rat Gene 1.0 ST Array 10857678 - -
          Affymetrix Rat Gene 1.0 ST Array 10857683 - -
          Affymetrix Rat Gene 1.0 ST Array 10857689 - -
          Affymetrix Rat Gene 1.1 ST Array 10857673 - -
          Rat Genome 230 2.0 Array 1378705_PM_at - -
          Rat Genome 230 2.0 Array 1394520_PM_at - -
          Rat Genome 230 2.0 Array 1391170_at - -
          Rat Genome 230 2.0 Array 1391170_PM_at - -
          Rat Genome 230 2.0 Array 1393317_PM_at - -
          Rat Genome 230 2.0 Array 1399070_PM_at - -
          Rat Genome 230 2.0 Array 1393317_at - -
          Rat Genome 230 2.0 Array 1378705_at - -
          Rat Genome 230 2.0 Array 1399070_at - -
          Rat Genome 230 2.0 Array 1394520_at - -
          Affymetrix HT Rat Focus Array 1399070_at - -
          Affymetrix HT Rat Focus Array 1378705_PM_at - -
          Affymetrix HT Rat Focus Array 1378705_at - -
          Affymetrix HT Rat Focus Array 1391170_at - -
          Affymetrix HT Rat Focus Array 1391170_PM_at - -
          Affymetrix HT Rat Focus Array 1399070_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400010940.rn.1 TC0400001563.rn.1 RTA TC
          Rat Expression Set 230 1399070_PM_at - -
          Rat Expression Set 230 1399070_at - -
          Rat Expression Set 230 1378705_at - -
          Rat Expression Set 230 1391170_PM_at - -
          Rat Expression Set 230 1391170_at - -
          Rat Expression Set 230 1394520_PM_at - -
          Rat Expression Set 230 1378705_PM_at - -
          Rat Expression Set 230 1394520_at - -
          Affymetrix Rat Gene 1.0 ST Array 10857673 - -
          Rat Genome U34 Set rc_AI180081_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21447821 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369675 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline