Search
Search
| Gene Symbol: Fbln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006236922.2 | XP_006236984.1 | 4 - 5 | 1809 | 66 | |
| XM_006236923.2 | XP_006236985.1 | 4 - 5 | 1621 | 66 | ||
| XM_006236924.2 | XP_006236986.1 | 4 - 5 | 1809 | 66 | ||
| XM_008763139.1 | XP_008761361.1 | 4 - 5 | 1555 | 66 | ||
Set Membership: |
|