Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01426179_g1
          See other SEC13 GE Assays ›
          Gene Symbol:
          Sec13
          Gene Name:
          SEC13 homolog (S. cerevisiae)
          Gene Aliases:
          Sec13l1
          Chromosome Location:
          Chr.4: 145686186 - 145699577 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01426179_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sec13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001006978.1 NP_001006979.1 6 - 7 610 61
          GenBank mRNA BC084705.1 - 6 - 7 610 7301091 61
          FQ212944.1 - 6 - 7 630 61
          FQ216715.1 - 6 - 7 628 61

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sec13

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s)

          ER to Golgi vesicle-mediated transport
          protein transport
          mRNA transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P832028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36483 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15928 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176498 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P832028 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P832028 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P832028 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P832028 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22342 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27451 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10864683 - -
          Affymetrix Rat Gene 1.0 ST Array 10864682 - -
          Affymetrix Rat Gene 1.0 ST Array 10864681 - -
          Affymetrix Rat Gene 1.1 ST Array 10864680 - -
          Rat Genome 230 2.0 Array 1371437_at - -
          Rat Genome 230 2.0 Array 1371437_PM_at - -
          Affymetrix HT Rat Focus Array 1371437_PM_at - -
          Affymetrix HT Rat Focus Array 1371437_at - -
          Affymetrix Rat Clariom S Assay PSR0400024910.rn.1 TC0400003551.rn.1 RTA TC
          Rat Expression Set 230 1371437_at - -
          Rat Expression Set 230 1371437_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10864680 - -
          Rat Genome U34 Set rc_AI232083_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22029224 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351124 - -
          RatRef-12 Expression BeadChip ILMN_70390 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline