Search
Search

RGD1559896| Gene Symbol: RGD1559896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109134.1 | NP_001102604.1 | 2 - 3 | 435 | 72 | |
| XM_008772642.1 | XP_008770864.1 | 2 - 3 | 452 | 72 | ||
| XM_008772643.1 | XP_008770865.1 | 2 - 3 | 335 | 72 | ||
| GenBank mRNA | FQ212098.1 | - | 2 - 3 | 177 | 72 | |
Set Membership: |
|