Search
Search
| Gene Symbol: Sprtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106199.1 | NP_001099669.1 | 3 - 4 | 601 | 85 | |
| XM_006255808.2 | XP_006255870.1 | 4 - 5 | 993 | 85 | ||
| XM_006255809.2 | XP_006255871.1 | 4 - 5 | 750 | 85 | ||
| XM_006255810.2 | XP_006255872.1 | 4 - 5 | 776 | 85 | ||
Set Membership: |
|