Search
Search
| Gene Symbol: Ccsap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107441.1 | NP_001100911.1 | 1 - 2 | 402 | 63 | |
| XM_006255811.2 | XP_006255873.1 | 2 - 3 | 902 | 63 | ||
| GenBank mRNA | FQ211944.1 | - | 1 - 2 | 176 | 63 | |
| FQ212467.1 | - | 1 - 2 | 62 | |||
Set Membership: |
|