Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01426651_m1
          See other RGD1562218 GE Assays ›
          Gene Symbol:
          RGD1562218
          Gene Name:
          similar to RIKEN cDNA 0610039J04
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 57600231 - 57614558 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01426651_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1562218
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034919.1 NP_001030091.1 5 - 6 663 88
          GenBank mRNA BC105747.1 - 5 - 6 663 5597965 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001691.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RGD1562218

          Gene Ontology Categories:

          Process(es)

          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P129652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824758 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86271 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129652 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129652 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P129652 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P257995 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67059 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67471 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10811946 - -
          Affymetrix Rat Gene 1.0 ST Array 10811941 - -
          Affymetrix Rat Gene 1.0 ST Array 10811945 - -
          Affymetrix Rat Gene 1.0 ST Array 10811944 - -
          Affymetrix Rat Gene 1.0 ST Array 10811940 - -
          Affymetrix Rat Gene 1.0 ST Array 10811943 - -
          Affymetrix Rat Gene 1.0 ST Array 10811942 - -
          Affymetrix Rat Gene 1.1 ST Array 10811939 - -
          Rat Genome 230 2.0 Array 1398136_at - -
          Rat Genome 230 2.0 Array 1389399_at - -
          Rat Genome 230 2.0 Array 1377838_PM_at - -
          Rat Genome 230 2.0 Array 1377838_at - -
          Rat Genome 230 2.0 Array 1389399_PM_at - -
          Rat Genome 230 2.0 Array 1398136_PM_at - -
          Affymetrix HT Rat Focus Array 1377838_PM_at - -
          Affymetrix HT Rat Focus Array 1398136_PM_at - -
          Affymetrix HT Rat Focus Array 1398136_at - -
          Affymetrix HT Rat Focus Array 1389399_at - -
          Affymetrix HT Rat Focus Array 1377838_at - -
          Affymetrix HT Rat Focus Array 1389399_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900013637.rn.1 TC1900001691.rn.1 RTA TC
          Rat Expression Set 230 1389399_PM_at - -
          Rat Expression Set 230 1389399_at - -
          Rat Expression Set 230 1398136_PM_at - -
          Rat Expression Set 230 1377838_PM_at - -
          Rat Expression Set 230 1377838_at - -
          Rat Expression Set 230 1398136_at - -
          Affymetrix Rat Gene 1.0 ST Array 10811939 - -
          Rat Genome U34 Set rc_AI058856_at - -
          Rat Genome U34 Set rc_AI236065_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21339694 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352045 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline