Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01426696_m1
          See other ARV1 GE Assays ›
          Gene Symbol:
          Arv1
          Gene Name:
          ARV1 homolog (S. cerevisiae)
          Gene Aliases:
          RGD1310264
          Chromosome Location:
          Chr.19: 57484720 - 57496539 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01426696_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arv1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106197.1 NP_001099667.1 4 - 5 709 60
          XM_006255806.2 XP_006255868.1 4 - 5 667 60
          XM_008772606.1 XP_008770828.1 4 - 5 620 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000837.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          bile acid metabolic process
          cholesterol transport
          regulation of cholesterol metabolic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P384645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88011 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14657 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P384645 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P384645 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P384645 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P384645 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58381 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43565 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10808837 - -
          Affymetrix Rat Gene 1.0 ST Array 10808839 - -
          Affymetrix Rat Gene 1.0 ST Array 10808840 - -
          Affymetrix Rat Gene 1.0 ST Array 10808842 - -
          Affymetrix Rat Gene 1.0 ST Array 10808838 - -
          Affymetrix Rat Gene 1.0 ST Array 10808841 - -
          Affymetrix Rat Gene 1.1 ST Array 10808836 - -
          Rat Genome 230 2.0 Array 1389057_PM_at - -
          Rat Genome 230 2.0 Array 1389057_at - -
          Affymetrix HT Rat Focus Array 1389057_at - -
          Affymetrix HT Rat Focus Array 1389057_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900006877.rn.1 TC1900000837.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900006881.rn.1 TC1900000837.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900006878.rn.1 TC1900000837.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900006880.rn.1 TC1900000837.rn.1 RTA TC
          Rat Expression Set 230 1389057_PM_at - -
          Rat Expression Set 230 1389057_at - -
          Affymetrix Rat Gene 1.0 ST Array 10808836 - -
          Rat Genome U34 Set rc_AA848503_g_at - -
          Rat Genome U34 Set rc_AA848503_at - -
          Rat Genome U34 Set rc_AA858503_at - -
          Rat Genome U34 Set H32451_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21559413 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371992 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0